[4-(2-Oxoimidazolidin-1-yl)phenyl]acetic acid 98%
|
|
BLDPBD24218-1GEA
6870
CZK
BLDPBD24218-1G
BLDPBD24218-100MG
BLDPBD24218-250MG
[4-(2-Oxoimidazolidin-1-yl)phenyl]acetic acid 98%
[4-(2-Oxoimidazolidin-1-yl)phenyl]acetic acid
Storage condition: Sealed in dry, Room Temperature
Vzorec:
C₁₁H₁₂N₂O₃ MW: 220,225 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD05864552 CAS číslo: 492445-92-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...