4-Iodo-2-methoxyphenol 97%
|
|
BLDPBD242434-100MGEA
513
CZK
BLDPBD242434-100MG
BLDPBD242434-250MG
BLDPBD242434-5G
BLDPBD242434-1G
4-Iodo-2-methoxyphenol 97%
4-Iodo-2-methoxyphenol
Storage condition: Keep in dark place, Sealed in dry, 2-8°C
Vzorec:
C₇H₇IO₂ MW: 250,034 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD08061924 CAS číslo: 203861-62-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...