2-Aminoimidazole hemisulphate 97%
|
|
BLDPBD242742-10GEA
371
CZK
BLDPBD242742-10G
BLDPBD242742-5G
BLDPBD242742-0.5G
BLDPBD242742-1G
BLDPBD242742-100G
BLDPBD242742-500G
BLDPBD242742-25G
2-Aminoimidazole hemisulphate 97%
2-Aminoimidazole hemisulphate
Storage condition: Inert atmosphere, Room Temperature
Vzorec:
C₆H₁₂N₆O₄S MW: 264,26 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00013162 CAS číslo: 1450-93-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...