Tisk…

4-Amino-2-methylbenzyl Alcohol 97%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD243570-5GEA 37370 CZK
BLDPBD243570-5G BLDPBD243570-1G BLDPBD243570-250MG BLDPBD243570-100MG BLDPBD243570-10G
4-Amino-2-methylbenzyl Alcohol 97%
4-Amino-2-methylbenzyl Alcohol

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Vzorec: C₈H₁₁NO
MW: 137,179 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11110504
CAS číslo: 63405-88-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR