Tisk…

4-Bromo-2-nitrophenylboronic acid 95%

BLDPBD243676-5GEA 22180 CZK
BLDPBD243676-5G BLDPBD243676-250MG BLDPBD243676-1G BLDPBD243676-100MG
4-Bromo-2-nitrophenylboronic acid 95%
4-Bromo-2-nitrophenylboronic acid

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₆H₅BBrNO₄
MW: 245,8232 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12025992
CAS číslo: 860034-11-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR