Tisk…

4-Methoxybenzo[d]oxazole-2(3H)-thione 95%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD248868-250MGEA 1400 CZK
BLDPBD248868-250MG BLDPBD248868-1G BLDPBD248868-100MG
4-Methoxybenzo[d]oxazole-2(3H)-thione 95%
4-Methoxybenzo[d]oxazole-2(3H)-thione

Storage condition: Sealed in dry, 2-8°C

Vzorec: C₈H₇NO₂S
MW: 181,212 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD17171374
CAS číslo: 1246471-39-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR