Tisk…

5-Chloroisoquinolin-1(2H)-one 95%

BLDPBD248919-250MGEA 2230 CZK
BLDPBD248919-250MG BLDPBD248919-100MG BLDPBD248919-1G
5-Chloroisoquinolin-1(2H)-one 95%
5-Chloroisoquinolin-1(2H)-one

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₉H₆ClNO
MW: 179,603 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11846293
CAS číslo: 24188-73-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR