Tisk…

3-(3'-Fluorobenzyloxy)phenylboronic acid 98%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD250393-1GEA 1540 CZK
BLDPBD250393-1G BLDPBD250393-100MG BLDPBD250393-5G BLDPBD250393-250MG
3-(3'-Fluorobenzyloxy)phenylboronic acid 98%
3-(3'-Fluorobenzyloxy)phenylboronic acid

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Vzorec: C₁₃H₁₂BFO₃
MW: 246,0420 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08276822
CAS číslo: 1072951-62-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR