Tisk…

2-Cyano-6-fluorophenylboronic acid 95%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD250404-250MGEA 1290 CZK
BLDPBD250404-250MG BLDPBD250404-1G BLDPBD250404-100MG
2-Cyano-6-fluorophenylboronic acid 95%
2-Cyano-6-fluorophenylboronic acid

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₇H₅BFNO₂
MW: 164,9295 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09033623
CAS číslo: 656235-44-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR