Tisk…

6-Formylpyridine-3-boronic acid, pinacol ester 98%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD25238-100MGEA 2170 CZK
BLDPBD25238-100MG
6-Formylpyridine-3-boronic acid, pinacol ester 98%
6-Formylpyridine-3-boronic acid, pinacol ester

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₁₂H₁₆BNO₃
MW: 233,071 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06659508
CAS číslo: 1073354-14-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR