Tisk…

3-Amino-5-bromo-2-methoxypyridine 98%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD25474-5GEA 542 CZK
BLDPBD25474-5G BLDPBD25474-25G BLDPBD25474-1G BLDPBD25474-100G BLDPBD25474-10G
3-Amino-5-bromo-2-methoxypyridine 98%
3-Amino-5-bromo-2-methoxypyridine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Vzorec: C₆H₇BrN₂O
MW: 203,04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07368887
CAS číslo: 884495-39-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR