Tisk…

(1R,5S,6R)-(3-Azabicyclo[3.1.0]hex-6-yl)methanol 95%

BLDPBD256844-100MGEA 1460 CZK
BLDPBD256844-100MG BLDPBD256844-5G BLDPBD256844-1G BLDPBD256844-250MG
(1R,5S,6R)-(3-Azabicyclo[3.1.0]hex-6-yl)methanol 95%
(1R,5S,6R)-(3-Azabicyclo[3.1.0]hex-6-yl)methanol

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Vzorec: C₆H₁₁NO
MW: 113,158 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11099879
CAS číslo: 134575-13-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR