Tisk…

(4-Biphenylylmethyl)hydrazine hydrochloride 95%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD257714-100MGEA 1520 CZK
BLDPBD257714-100MG BLDPBD257714-250MG BLDPBD257714-1G
(4-Biphenylylmethyl)hydrazine hydrochloride 95%
(4-Biphenylylmethyl)hydrazine hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₁₃H₁₄N₂·ClH
MW: 234,725 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01722362
CAS číslo: 75333-07-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR