3-Bromo-5-(trifluoromethoxy)benzonitrile 98+%
|
|
BLDPBD257987-250MGEA
627
CZK
BLDPBD257987-250MG
BLDPBD257987-5G
BLDPBD257987-100MG
BLDPBD257987-1G
3-Bromo-5-(trifluoromethoxy)benzonitrile 98+%
3-Bromo-5-(trifluoromethoxy)benzonitrile
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Vzorec:
C₈H₃BrF₃NO MW: 266,01 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08458074 CAS číslo: 914635-52-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...