Tisk…

3-Bromo-1-methyl-1H-indazole 98%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD258498-1GEA 2340 CZK
BLDPBD258498-1G BLDPBD258498-5G BLDPBD258498-250MG
3-Bromo-1-methyl-1H-indazole 98%
3-Bromo-1-methyl-1H-indazole

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Vzorec: C₈H₇BrN₂
MW: 211,0586 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12923368
CAS číslo: 326474-67-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR