Tisk…

(S)-1-(3-Ethoxy-4-methoxyphenyl)-2-(methylsulphonyl)ethanamine 98%

BLDPBD258927-5GEA 1060 CZK
BLDPBD258927-5G BLDPBD258927-25G BLDPBD258927-1G BLDPBD258927-250MG
(S)-1-(3-Ethoxy-4-methoxyphenyl)-2-(methylsulphonyl)ethanamine 98%
(S)-1-(3-Ethoxy-4-methoxyphenyl)-2-(methylsulphonyl)ethanamine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Vzorec: C₁₂H₁₉NO₄S
MW: 273,349 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22677268
CAS číslo: 608141-42-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR