Tisk…

a-Cyclopropyl-4-chloro-benzylamine 95%

BLDPBD259036-100MGEA 3680 CZK
BLDPBD259036-100MG BLDPBD259036-1G BLDPBD259036-250MG
a-Cyclopropyl-4-chloro-benzylamine 95%
a-Cyclopropyl-4-chloro-benzylamine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Vzorec: C₁₀H₁₂ClN
MW: 181,662 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05191965
CAS číslo: 123312-22-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR