Tisk…

Methyl-8-bromoimidazo[1,2-a]pyridine-6-carboxylate 96%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD259037-250MGEA 1090 CZK
BLDPBD259037-250MG BLDPBD259037-1G
Methyl-8-bromoimidazo[1,2-a]pyridine-6-carboxylate 96%
Methyl-8-bromoimidazo[1,2-a]pyridine-6-carboxylate

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Vzorec: C₉H₇BrN₂O₂
MW: 255,0681 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD17016026
CAS číslo: 1234616-08-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR