Tisk…

6-Chloro-5-methyl-1H-indazole 98%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD260896-250MGEA 1090 CZK
BLDPBD260896-250MG BLDPBD260896-1G BLDPBD260896-100MG BLDPBD260896-5G
6-Chloro-5-methyl-1H-indazole 98%
6-Chloro-5-methyl-1H-indazole

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₈H₇ClN₂
MW: 166,608 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09880179
CAS číslo: 1000341-02-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR