Tisk…

2-(3'-Chlorobenzyloxy)phenylboronic acid 98%

BLDPBD260919-1GEA 4540 CZK
BLDPBD260919-1G BLDPBD260919-100MG BLDPBD260919-250MG
2-(3'-Chlorobenzyloxy)phenylboronic acid 98%
2-(3'-Chlorobenzyloxy)phenylboronic acid

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₁₃H₁₂BClO₃
MW: 262,5000 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06411351
CAS číslo: 849062-32-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR