Tisk…

4-Amino-3-cyanobenzoic acid 95%

BLDPBD262097-250MGEA 1520 CZK
BLDPBD262097-250MG BLDPBD262097-1G BLDPBD262097-10G BLDPBD262097-100MG BLDPBD262097-5G
4-Amino-3-cyanobenzoic acid 95%
4-Amino-3-cyanobenzoic acid

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Vzorec: C₈H₆N₂O₂
MW: 162,145 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09745178
CAS číslo: 74192-47-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR