Tisk…

(1-Phenyl-1H-1,2,3-triazol-4-yl)methanol 95%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD262863-1GEA 2660 CZK
BLDPBD262863-1G BLDPBD262863-250MG
(1-Phenyl-1H-1,2,3-triazol-4-yl)methanol 95%
(1-Phenyl-1H-1,2,3-triazol-4-yl)methanol

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₉H₉N₃O
MW: 175,1900 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00100214
CAS číslo: 103755-58-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR