Tisk…

6-(Methoxycarbonyl)indole-2-boronic acid pinacol ester 96%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD265200-5GEA 16910 CZK
BLDPBD265200-5G BLDPBD265200-100MG BLDPBD265200-250MG BLDPBD265200-1G
6-(Methoxycarbonyl)indole-2-boronic acid pinacol ester 96%
6-(Methoxycarbonyl)indole-2-boronic acid pinacol ester

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₁₆H₂₀BNO₄
MW: 301,1453 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11858368
CAS číslo: 1256359-21-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR