Tisk…

3-Bromo-2-nitrobenzaldehyde 97%

BLDPBD266813-100MGEA 1170 CZK
BLDPBD266813-100MG BLDPBD266813-1G BLDPBD266813-250MG
3-Bromo-2-nitrobenzaldehyde 97%
3-Bromo-2-nitrobenzaldehyde
Vzorec: C₇H₄BrNO₃
MW: 230,016 g/mol
MDL Number: MFCD00462872
CAS číslo: 882722-99-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR