Tisk…

Tri-tert-butylmethanol 98%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD268405-1GEA 12570 CZK
BLDPBD268405-1G BLDPBD268405-250MG BLDPBD268405-100MG
Tri-tert-butylmethanol 98%
Tri-tert-butylmethanol

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₁₃H₂₈O
MW: 200,361 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00204381
CAS číslo: 41902-42-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR