Tisk…

4-Cyano-3-fluorobenzaldehyde 97%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD27110-1GEA 713 CZK
BLDPBD27110-1G BLDPBD27110-25G BLDPBD27110-250MG BLDPBD27110-10G BLDPBD27110-5G
4-Cyano-3-fluorobenzaldehyde 97%
4-Cyano-3-fluorobenzaldehyde

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₈H₄FNO
MW: 149,1200 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06738989
CAS číslo: 101048-76-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR