Tisk…

2-Amino-4-methylthiophene-3-carbonitrile 98%

BLDPBD271558-5GEA 2000 CZK
BLDPBD271558-5G BLDPBD271558-1G
2-Amino-4-methylthiophene-3-carbonitrile 98%
2-Amino-4-methylthiophene-3-carbonitrile

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Vzorec: C₆H₆N₂S
MW: 138,19 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00277568
CAS číslo: 4623-55-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR