Tisk…

Methyl 4-chloro-3-cyanobenzoate 98%

BLDPBD272407-250MGEA 1460 CZK
BLDPBD272407-250MG
Methyl 4-chloro-3-cyanobenzoate 98%
Methyl 4-chloro-3-cyanobenzoate

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₉H₆ClNO₂
MW: 195,602 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18398898
CAS číslo: 181282-80-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR