Tisk…

3-Bromo-5-methylimidazo[1,2-a]pyridine 98%

BLDPBD272746-5GEA 9500 CZK
BLDPBD272746-5G BLDPBD272746-1G BLDPBD272746-250MG
3-Bromo-5-methylimidazo[1,2-a]pyridine 98%
3-Bromo-5-methylimidazo[1,2-a]pyridine

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₈H₇BrN₂
MW: 211,059 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD19441944
CAS číslo: 5857-47-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR