Tisk…

5-Methyl-2-(phenylethynyl)pyridine 95%

BLDPBD273431-250MGEA 2280 CZK
BLDPBD273431-250MG BLDPBD273431-1G BLDPBD273431-100MG
5-Methyl-2-(phenylethynyl)pyridine 95%
5-Methyl-2-(phenylethynyl)pyridine

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₁₄H₁₁N
MW: 193,244 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18205120
CAS číslo: 124300-38-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR