Ethyl-2-aminoisonicotinate 98%
|
|
BLDPBD27450-250MGEA
171
CZK
BLDPBD27450-250MG
BLDPBD27450-100G
BLDPBD27450-10G
BLDPBD27450-1G
BLDPBD27450-25G
BLDPBD27450-500G
BLDPBD27450-5G
Ethyl-2-aminoisonicotinate 98%
Ethyl-2-aminoisonicotinate
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Vzorec:
C₈H₁₀N₂O₂ MW: 166,18 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03791260 CAS číslo: 13362-30-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...