4-Amino-5-bromopyrimidine 98%
|
|
BLDPBD27527-10GEA
10690
CZK
BLDPBD27527-10G
BLDPBD27527-1G
BLDPBD27527-5G
BLDPBD27527-250MG
BLDPBD27527-25G
4-Amino-5-bromopyrimidine 98%
4-Amino-5-bromopyrimidine
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Vzorec:
C₄H₄BrN₃ MW: 174,00 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00233962 CAS číslo: 1439-10-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...