Tisk…

5-Phenyl-3-isoxazolecarboxylic acid 97%

BLDPBD27536-250MGEA 200 CZK
BLDPBD27536-250MG BLDPBD27536-25G BLDPBD27536-10G BLDPBD27536-5G BLDPBD27536-1G BLDPBD27536-100MG
5-Phenyl-3-isoxazolecarboxylic acid 97%
5-Phenyl-3-isoxazolecarboxylic acid

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Vzorec: C₁₀H₇NO₃
MW: 189,1700 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00464221
CAS číslo: 14441-90-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR