Tisk…

5-Methyl-1H-indazole 97%

BLDPBD27808-250MGEA 228 CZK
BLDPBD27808-250MG BLDPBD27808-1G BLDPBD27808-5G BLDPBD27808-10G
5-Methyl-1H-indazole 97%
5-Methyl-1H-indazole

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₈H₈N₂
MW: 132,1600 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01606235
CAS číslo: 1776-37-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR