Tisk…

Ethyl-2-amino-4-oxazolecarboxylate 98%

BLDPBD27810-10GEA 1140 CZK
BLDPBD27810-10G BLDPBD27810-25G BLDPBD27810-100G BLDPBD27810-5G BLDPBD27810-1G BLDPBD27810-250MG
Ethyl-2-amino-4-oxazolecarboxylate 98%
Ethyl-2-amino-4-oxazolecarboxylate

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Vzorec: C₆H₈N₂O₃
MW: 156,14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06659907
CAS číslo: 177760-52-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR