Tisk…

5-Bromo-2-chlorobenzamide 96%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD278634-250MGEA 941 CZK
BLDPBD278634-250MG BLDPBD278634-5G BLDPBD278634-1G
5-Bromo-2-chlorobenzamide 96%
5-Bromo-2-chlorobenzamide

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₇H₅BrClNO
MW: 234,4777 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00482914
CAS číslo: 188774-55-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR