Tisk…

6-Bromo-1-chloroisoquinoline 97%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD27983-5GEA 4170 CZK
BLDPBD27983-5G BLDPBD27983-10G BLDPBD27983-100G BLDPBD27983-1G BLDPBD27983-250MG BLDPBD27983-25G
6-Bromo-1-chloroisoquinoline 97%
6-Bromo-1-chloroisoquinoline

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₉H₅BrClN
MW: 242,5000 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06738661
CAS číslo: 205055-63-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR