Tisk…

Ethyl-6-fluoroimidazo[1,2-a]pyridine-3-carboxylate 97%

BLDPBD280375-1GEA 1400 CZK
BLDPBD280375-1G BLDPBD280375-5G BLDPBD280375-250MG
Ethyl-6-fluoroimidazo[1,2-a]pyridine-3-carboxylate 97%
Ethyl-6-fluoroimidazo[1,2-a]pyridine-3-carboxylate

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₁₀H₉FN₂O₂
MW: 208,1900 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21364586
CAS číslo: 1359655-87-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR