Tisk…

Methyl-2-bromoisonicotinate 97%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD28208-5GEA 770 CZK
BLDPBD28208-5G BLDPBD28208-1G BLDPBD28208-100G BLDPBD28208-10G BLDPBD28208-25G
Methyl-2-bromoisonicotinate 97%
Methyl-2-bromoisonicotinate

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Vzorec: C₇H₆BrNO₂
MW: 216,0320 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03791265
CAS číslo: 26156-48-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR