Tisk…

7-Bromo-6-methyl-1H-indazole 97%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD283209-5GEA 33920 CZK
BLDPBD283209-5G BLDPBD283209-100MG BLDPBD283209-1G BLDPBD283209-250MG
7-Bromo-6-methyl-1H-indazole 97%
7-Bromo-6-methyl-1H-indazole

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Vzorec: C₈H₇BrN₂
MW: 211,059 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD17168273
CAS číslo: 1257535-45-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR