Tisk…

Isopropyl 2-acetamido-2-deoxy-3,4,6-tri-O-acetyl-α-D-glucopyranoside 98%

BLDPBD284755-250MGEA 19210 CZK
BLDPBD284755-250MG BLDPBD284755-10MG BLDPBD284755-100MG BLDPBD284755-50MG BLDPBD284755-25MG
Isopropyl 2-acetamido-2-deoxy-3,4,6-tri-O-acetyl-α-D-glucopyranoside 98%
Isopropyl 2-acetamido-2-deoxy-3,4,6-tri-O-acetyl-α-D-glucopyranoside

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Vzorec: C₁₇H₂₇NO₉
MW: 389,398 g/mol
Storage Temperature: Freezer
MDL Number: MFCD28053519
CAS číslo: 40592-88-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR