Tisk…

2-Chloro-3-(trichloromethyl)pyridine 97%

BLDPBD285003-250MGEA 371 CZK
BLDPBD285003-250MG BLDPBD285003-1G
2-Chloro-3-(trichloromethyl)pyridine 97%
2-Chloro-3-(trichloromethyl)pyridine

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₆H₃Cl₄N
MW: 230,907 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00160150
CAS číslo: 72648-12-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR