Tisk…

4-Methoxy-2-nitrobenzonitrile 95%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD28567-25GEA 5790 CZK
BLDPBD28567-25G BLDPBD28567-5G BLDPBD28567-0.025G BLDPBD28567-1G BLDPBD28567-10G
4-Methoxy-2-nitrobenzonitrile 95%
4-Methoxy-2-nitrobenzonitrile

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₈H₆N₂O₃
MW: 178,1500 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00667741
CAS číslo: 38469-83-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR