Tisk…

1-(3-Bromophenyl)cyclopropan-1-amine hydrochloride 98%

BLDPBD290855-5GEA 7760 CZK
BLDPBD290855-5G BLDPBD290855-250MG BLDPBD290855-1G
1-(3-Bromophenyl)cyclopropan-1-amine hydrochloride 98%
1-(3-Bromophenyl)cyclopropan-1-amine hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₉H₁₀BrN·ClH
MW: 248,547 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11616353
CAS číslo: 597563-15-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR