Tisk…

8-Isoquinolinecarboxylic acid 95%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD29231-250MGEA 656 CZK
BLDPBD29231-250MG BLDPBD29231-5G BLDPBD29231-25G BLDPBD29231-100MG BLDPBD29231-10G BLDPBD29231-1G
8-Isoquinolinecarboxylic acid 95%
8-Isoquinolinecarboxylic acid

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₁₀H₇NO₂
MW: 173,1681 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06738935
CAS číslo: 61563-43-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR