Tisk…

1,6-Dichloroisoquinoline 97%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD29274-1GEA 2310 CZK
BLDPBD29274-1G BLDPBD29274-5G BLDPBD29274-250MG BLDPBD29274-100MG
1,6-Dichloroisoquinoline 97%
1,6-Dichloroisoquinoline

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₉H₅Cl₂N
MW: 198,0500 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06738660
CAS číslo: 630421-73-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR