Tisk…

Imidazo[1,2-a]pyridine-2-carboxylic acid 98%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD29316-1GEA 941 CZK
BLDPBD29316-1G BLDPBD29316-25G BLDPBD29316-250MG BLDPBD29316-5G
Imidazo[1,2-a]pyridine-2-carboxylic acid 98%
Imidazo[1,2-a]pyridine-2-carboxylic acid

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₈H₆N₂O₂
MW: 162,1500 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03419462
CAS číslo: 64951-08-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR