Tisk…

1-Benzyl-4-chloropiperidine 95%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD29399-5GEA 14880 CZK
BLDPBD29399-5G BLDPBD29399-1G BLDPBD29399-250MG
1-Benzyl-4-chloropiperidine 95%
1-Benzyl-4-chloropiperidine

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₁₂H₁₆ClN
MW: 209,715 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00510091
CAS číslo: 67848-71-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR