5-Quinoxalinecarboxylic acid 97%
|
|
BLDPBD29433-10GEA
13570
CZK
BLDPBD29433-10G
BLDPBD29433-5G
BLDPBD29433-1G
BLDPBD29433-0.025G
BLDPBD29433-25G
BLDPBD29433-250MG
5-Quinoxalinecarboxylic acid 97%
5-Quinoxalinecarboxylic acid
Storage condition: Sealed in dry, Room Temperature
Vzorec:
C₉H₆N₂O₂ MW: 174,1561 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02854494 CAS číslo: 6924-66-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...