Tisk…

2,3-Diamino-6-chloro-benzonitrile 95%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD294419-250MGEA 399 CZK
BLDPBD294419-250MG BLDPBD294419-5G BLDPBD294419-1G
2,3-Diamino-6-chloro-benzonitrile 95%
2,3-Diamino-6-chloro-benzonitrile

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Vzorec: C₇H₆ClN₃
MW: 167,596 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD20723943
CAS číslo: 548457-80-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR